Product Details

SNP ID
rs139503409
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15496583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAATGTCAATTGTAGAATCCAGA[C/T]GGTGGTGTATTGGTGCTTTAACTTT
Phenotype
MIM: 300335 MIM: 300101 MIM: 300931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACE2 PubMed Links

Gene Details

Gene
ACE2
Gene Name
angiotensin I converting enzyme 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021804.2 Intron NP_068576.1
XM_011545549.1 Intron XP_011543851.1
XM_011545551.2 Intron XP_011543853.1
XM_011545552.1 Intron XP_011543854.1
Gene
BMX
Gene Name
BMX non-receptor tyrosine kinase
There are no transcripts associated with this gene.

Gene
PIR
Gene Name
pirin
There are no transcripts associated with this gene.

Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details