Product Details

SNP ID
rs139605296
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:152736574 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCACAAAGGCAGAAATGCTGGGGA[G/C]TGTCATCAGAAATTTCCAGGACTTC
Phenotype
MIM: 300944 MIM: 300177
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
CSAG1 PubMed Links

Gene Details

Gene
CSAG1
Gene Name
chondrosarcoma associated gene 1
There are no transcripts associated with this gene.

Gene
CSAG4
Gene Name
CSAG family member 4 (pseudogene)
There are no transcripts associated with this gene.

Gene
MAGEA12
Gene Name
MAGE family member A12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166386.3 767 Missense Mutation ACT,AGT T138S NP_001159858.1
NM_001166387.3 767 Missense Mutation ACT,AGT T138S NP_001159859.1
NM_005367.6 767 Missense Mutation ACT,AGT T138S NP_005358.2

View Full Product Details