Product Details

SNP ID
rs139628291
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:49275994 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGGAGTTGAACGGGACAAGTCAG[A/G]GGCATCTTAGTTTTCTGTAGAGTGA
Phenotype
MIM: 300292
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FOXP3 PubMed Links

Gene Details

Gene
FOXP3
Gene Name
forkhead box P3
There are no transcripts associated with this gene.

Gene
PPP1R3F
Gene Name
protein phosphatase 1 regulatory subunit 3F
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184745.1 3925 Intron NP_001171674.1
NM_033215.4 3925 Intron NP_149992.3
XM_005272687.3 3925 Intron XP_005272744.1
XM_017029932.1 3925 UTR 5 XP_016885421.1
XM_017029933.1 3925 UTR 5 XP_016885422.1
XM_017029934.1 3925 Intron XP_016885423.1
XM_017029935.1 3925 Intron XP_016885424.1

View Full Product Details