Product Details

SNP ID
rs139841459
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153649266 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGATTCCAGGCCGAGTGCCCTCA[C/G]CTGTGTGAGACCAGCCTTGCTGGCC
Phenotype
MIM: 300134
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DUSP9 PubMed Links

Gene Details

Gene
DUSP9
Gene Name
dual specificity phosphatase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318503.1 525 Silent Mutation CAC,CAG H136Q NP_001305432.1
NM_001395.3 525 Silent Mutation CAC,CAG H136Q NP_001386.1
XM_011531123.1 525 Missense Mutation CAC,CAG H167Q XP_011529425.1
XM_011531124.1 525 Missense Mutation CAC,CAG H136Q XP_011529426.1

View Full Product Details