Product Details

SNP ID
rs139910673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30559921 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCATATTCAAAACTGCTCTCAGA[G/T]GGAAAATCTGTTGTCACTGAATTAA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 830 Silent Mutation CCA,CCC P145P NP_079435.1

View Full Product Details