Product Details

SNP ID
rs139974100
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137030571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGCTCTCCGGGATGTTCACTGC[C/T]TCGACGTCATCCTCACTGAAATTGA
Phenotype
MIM: 300393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR101 PubMed Links

Gene Details

Gene
GPR101
Gene Name
G protein-coupled receptor 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054021.1 1104 Silent Mutation GAA,GAG E368E NP_473362.1

View Full Product Details