Product Details

SNP ID
rs140025330
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:151404918 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACTGTTTTTTTTCTCTTGATAGGC[A/G]CCCTTGGGATGTCCAATAGGGACAG
Phenotype
MIM: 300913
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
VMA21 PubMed Links

Gene Details

Gene
VMA21
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017980.3 616 Missense Mutation ACC,GCC T56A NP_001017980.1
XM_011531125.2 616 Missense Mutation ACC,GCC T111A XP_011529427.1

View Full Product Details