Product Details

SNP ID
rs140112568
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48829477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCTGTCTGTGATGGTGTGCTGGG[C/T]GTCTTCGCTCTCGATGACCCCTCGT
Phenotype
MIM: 300437 MIM: 300272 MIM: 300399
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ERAS PubMed Links

Gene Details

Gene
ERAS
Gene Name
ES cell expressed Ras
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181532.3 434 Silent Mutation GGC,GGT G118G NP_853510.1
Gene
HDAC6
Gene Name
histone deacetylase 6
There are no transcripts associated with this gene.

Gene
PCSK1N
Gene Name
proprotein convertase subtilisin/kexin type 1 inhibitor
There are no transcripts associated with this gene.

View Full Product Details