Product Details
- SNP ID
-
rs140378313
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:136668145 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTTCTTGATTTCAGTTCTTCTTCCA[G/T]GCATTGCTTCATTCTTTTATTTTCC
- Phenotype
-
MIM: 300267
MIM: 300386
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
ARHGEF6
PubMed Links
Gene Details
- Gene
- ARHGEF6
- Gene Name
- Rac/Cdc42 guanine nucleotide exchange factor 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001306177.1 |
3082 |
Missense Mutation |
ATG,CTG |
M585L |
NP_001293106.1 |
NM_004840.2 |
3082 |
Missense Mutation |
ATG,CTG |
M739L |
NP_004831.1 |
XM_005262499.2 |
3082 |
Missense Mutation |
ATG,CTG |
M690L |
XP_005262556.1 |
XM_011531412.2 |
3082 |
Missense Mutation |
ATG,CTG |
M766L |
XP_011529714.1 |
XM_011531413.1 |
3082 |
Missense Mutation |
ATG,CTG |
M743L |
XP_011529715.1 |
XM_011531414.1 |
3082 |
Missense Mutation |
ATG,CTG |
M736L |
XP_011529716.1 |
XM_011531415.2 |
3082 |
Missense Mutation |
ATG,CTG |
M709L |
XP_011529717.1 |
XM_011531416.2 |
3082 |
Missense Mutation |
ATG,CTG |
M612L |
XP_011529718.1 |
XM_011531417.1 |
3082 |
Missense Mutation |
ATG,CTG |
M589L |
XP_011529719.1 |
XM_017029975.1 |
3082 |
Missense Mutation |
ATG,CTG |
M770L |
XP_016885464.1 |
XM_017029976.1 |
3082 |
Missense Mutation |
ATG,CTG |
M536L |
XP_016885465.1 |
- Gene
- CD40LG
- Gene Name
- CD40 ligand
There are no transcripts associated with this gene.
View Full Product Details