Product Details

SNP ID
rs140535851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:107524440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGACCTCATCCTAGCATCCCCTTC[C/T]ACCTCCCGAAGGGGTGACAGACAGA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FRMPD3 PubMed Links

Gene Details

Gene
FRMPD3
Gene Name
FERM and PDZ domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032428.1 Intron NP_115804.1
XM_006724709.2 Intron XP_006724772.1
XM_011531056.2 Intron XP_011529358.1
XM_011531057.2 Intron XP_011529359.1
XM_011531058.2 Intron XP_011529360.1
XM_017029901.1 Intron XP_016885390.1
XM_017029902.1 Intron XP_016885391.1
Gene
FRMPD3-AS1
Gene Name
FRMPD3 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details