Product Details

SNP ID
hCV175323007
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154013331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGGGCTCTGCCAGTGCTGGACA[C/T]GTAGGAGTTCTCCTGCGGGGAAGGG
Phenotype
MIM: 300283 MIM: 300005 MIM: 300929
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IRAK1 PubMed Links

Gene Details

Gene
IRAK1
Gene Name
interleukin 1 receptor associated kinase 1
There are no transcripts associated with this gene.

Gene
MECP2
Gene Name
methyl-CpG binding protein 2
There are no transcripts associated with this gene.

Gene
MIR718
Gene Name
microRNA 718
There are no transcripts associated with this gene.

View Full Product Details