Product Details
- SNP ID
-
rs140603041
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:100587278 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATATCATCTTTACATCTCATTCTAG[C/T]TCTGTTTTTAATGATTATAAGGAGC
- Phenotype
-
MIM: 300459
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TNMD
PubMed Links
Gene Details
- Gene
- TNMD
- Gene Name
- tenomodulin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022144.2 |
|
Intron |
|
|
NP_071427.2 |
View Full Product Details