Product Details

SNP ID
rs140665718
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:123605842 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAAACAAACAAAAAAAGGCTTAG[A/G]GCTGGAATTACAAAATCAATTGTCT
Phenotype
MIM: 300395
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THOC2 PubMed Links

Gene Details

Gene
THOC2
Gene Name
THO complex 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081550.1 Intron NP_001075019.1
XM_005262450.3 Intron XP_005262507.1
XM_017029662.1 Intron XP_016885151.1
XM_017029663.1 Intron XP_016885152.1
XM_017029664.1 Intron XP_016885153.1
XM_017029665.1 Intron XP_016885154.1
XM_017029666.1 Intron XP_016885155.1
XM_017029667.1 Intron XP_016885156.1
XM_017029668.1 Intron XP_016885157.1
XM_017029669.1 Intron XP_016885158.1
XM_017029670.1 Intron XP_016885159.1
XM_017029671.1 Intron XP_016885160.1
XM_017029672.1 Intron XP_016885161.1
XM_017029673.1 Intron XP_016885162.1
XM_017029674.1 Intron XP_016885163.1
XM_017029675.1 Intron XP_016885164.1
XM_017029676.1 Intron XP_016885165.1
XM_017029677.1 Intron XP_016885166.1
XM_017029678.1 Intron XP_016885167.1
XM_017029679.1 Intron XP_016885168.1

View Full Product Details