Product Details
- SNP ID
-
rs140818152
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:75783483 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGTGATACATATTCCCTAGACTTCA[C/T]GTGGGGTCAAGGAAAAACATGATAG
- Phenotype
-
MIM: 300760
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MAGEE2
PubMed Links
Gene Details
- Gene
- MAGEE2
- Gene Name
- MAGE family member E2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_138703.4 |
1762 |
Silent Mutation |
ACA,ACG |
T523T |
NP_619648.1 |
View Full Product Details