Product Details

SNP ID
rs140854514
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153420668 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGACAGAACACAGAGCAAGACGTC[A/G]ACTTCAACGCAGAAGCATTCTGGAC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZFP92 PubMed Links

Gene Details

Gene
ZFP92
Gene Name
ZFP92 zinc finger protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136273.1 552 Silent Mutation TCA,TCG S97S NP_001129745.1
XM_005274652.3 552 Silent Mutation TCA,TCG S97S XP_005274709.1
XM_011531115.2 552 Silent Mutation TCA,TCG S97S XP_011529417.1
XM_017029281.1 552 Silent Mutation TCA,TCG S193S XP_016884770.1

View Full Product Details