Product Details

SNP ID
rs140881112
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:104166803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCAACCATGTCCGACGAGGCCTC[C/G]GCCATCACTTCCTACGAGAAGTTTC
Phenotype
MIM: 300941
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM199X PubMed Links

Gene Details

Gene
FAM199X
Gene Name
family with sequence similarity 199, X-linked
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207318.3 326 Silent Mutation TCC,TCG S6S NP_997201.1
XM_005262079.2 326 Silent Mutation TCC,TCG S6S XP_005262136.1
XM_017029270.1 326 Intron XP_016884759.1
Gene
SLC25A53
Gene Name
solute carrier family 25 member 53
There are no transcripts associated with this gene.

View Full Product Details