Product Details

SNP ID
rs140975055
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:18252955 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCACGCTGAAATCTCTCACAGTCT[C/T]ATGATGCTTAAGAAATAAAGACCCA
Phenotype
MIM: 300208
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SCML2 PubMed Links

Gene Details

Gene
SCML2
Gene Name
sex comb on midleg-like 2 (Drosophila)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006089.2 Intron NP_006080.1
XM_006724458.1 Intron XP_006724521.1
XM_017029219.1 Intron XP_016884708.1
XM_017029220.1 Intron XP_016884709.1
XM_017029221.1 Intron XP_016884710.1
XM_017029222.1 Intron XP_016884711.1

View Full Product Details