Product Details

SNP ID
rs140989967
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81120677 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGTGTTATACGGTTTTACCAAGCA[A/G]TGAAACAGCGTTCGATTTCACTTCT
Phenotype
MIM: 300385
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030763.2 Intron NP_110390.1
XM_006724688.3 Intron XP_006724751.1
XM_017029839.1 Intron XP_016885328.1
XM_017029840.1 Intron XP_016885329.1

View Full Product Details