Product Details

SNP ID
rs141122494
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:50370538 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTGAGGCTTCTACTGTCAGTGTC[C/T]GAAGATTTTTCCTGAGAAACCACAA
Phenotype
MIM: 300837
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGKK PubMed Links

Gene Details

Gene
DGKK
Gene Name
diacylglycerol kinase kappa
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013742.3 3709 Silent Mutation TCA,TCG S1208S NP_001013764.1
XM_017029268.1 3709 Silent Mutation TCA,TCG S1179S XP_016884757.1

View Full Product Details