Product Details

SNP ID
rs141240078
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46574648 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGTGGCGATACGCGCCCTGGAGGC[A/C]GAGTGCCGAAAGTACCCGGTGGTGG
Phenotype
MIM: 300375
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019886.3 962 Silent Mutation GCA,GCC A239A NP_063939.2

View Full Product Details