Product Details

SNP ID
rs141266102
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:132214183 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTATCTTCTTTACTGGACGACACA[A/G]GTTGTACAACACTGATCTTGCTTCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RAP2C PubMed Links

Gene Details

Gene
RAP2C
Gene Name
RAP2C, member of RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271186.1 1461 Silent Mutation ACC,ACT T179T NP_001258115.1
NM_001271187.1 1461 Silent Mutation ACC,ACT T113T NP_001258116.1
NM_021183.4 1461 Silent Mutation ACC,ACT T179T NP_067006.3
XM_006724775.2 1461 Silent Mutation ACC,ACT T113T XP_006724838.1
XM_011531376.1 1461 Silent Mutation ACC,ACT T179T XP_011529678.1
Gene
RAP2C-AS1
Gene Name
RAP2C antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details