Product Details
- SNP ID
-
hCV175343992
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:48802784 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCCCTCAGGACTCCAGTGTCACTT[C/T]GGTGAGGCCCTAGACCCGCCCTGAT
- Phenotype
-
MIM: 305371
MIM: 300272
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GATA1
PubMed Links
Gene Details
- Gene
- GATA1
- Gene Name
- GATA binding protein 1
There are no transcripts associated with this gene.
- Gene
- HDAC6
- Gene Name
- histone deacetylase 6
There are no transcripts associated with this gene.
View Full Product Details