Product Details

SNP ID
rs141699493
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:149608820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACATGGCTTTAAACTCCACACAC[A/G]TTTCTCCTTCTGCTCTAAAAAAGGG
Phenotype
MIM: 300031
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM185A PubMed Links

Gene Details

Gene
TMEM185A
Gene Name
transmembrane protein 185A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001174092.2 385 Missense Mutation ACG,ATG T18M NP_001167563.1
NM_001282302.1 385 Missense Mutation ACG,ATG T77M NP_001269231.1
NM_032508.3 385 Missense Mutation ACG,ATG T77M NP_115897.1

View Full Product Details