Product Details

SNP ID
rs141752835
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:118746964 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTTTCTAACCTTAGAAAATAGCTC[C/T]GGAAACTCGTCGTTCAATAGAAGTA
Phenotype
MIM: 300119
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL13RA1 PubMed Links

Gene Details

Gene
IL13RA1
Gene Name
interleukin 13 receptor subunit alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001560.2 560 Missense Mutation CCG,CTG P80L NP_001551.1
XM_011531336.1 560 Missense Mutation CCG,CTG P182L XP_011529638.1
XM_011531337.1 560 Intron XP_011529639.1
XM_017029507.1 560 Intron XP_016884996.1

View Full Product Details