Product Details

SNP ID
rs142057050
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:136665967 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAAAGTATTCCGTAATCACAAAA[A/G]TGTTTCCTATAGATGCAGTTTGCAC
Phenotype
MIM: 300267 MIM: 300386
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF6 PubMed Links

Gene Details

Gene
ARHGEF6
Gene Name
Rac/Cdc42 guanine nucleotide exchange factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306177.1 5260 UTR 3 NP_001293106.1
NM_004840.2 5260 UTR 3 NP_004831.1
XM_005262499.2 5260 UTR 3 XP_005262556.1
XM_011531412.2 5260 UTR 3 XP_011529714.1
XM_011531413.1 5260 UTR 3 XP_011529715.1
XM_011531414.1 5260 UTR 3 XP_011529716.1
XM_011531415.2 5260 UTR 3 XP_011529717.1
XM_011531416.2 5260 UTR 3 XP_011529718.1
XM_011531417.1 5260 UTR 3 XP_011529719.1
XM_017029975.1 5260 UTR 3 XP_016885464.1
XM_017029976.1 5260 UTR 3 XP_016885465.1
Gene
CD40LG
Gene Name
CD40 ligand
There are no transcripts associated with this gene.

View Full Product Details