Product Details

SNP ID
rs142180569
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1465637 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCTGGGCACCCGGCGGCAGCCCA[A/G]ATATTCCCGCAGGCGCAGCTGGAAT
Phenotype
MIM: 300525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
P2RY8 PubMed Links

Gene Details

Gene
P2RY8
Gene Name
purinergic receptor P2Y8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178129.4 1287 Silent Mutation CTG,TTG L308L NP_835230.1
XM_005274429.3 1287 Silent Mutation CTG,TTG L308L XP_005274486.1
XM_006724443.3 1287 Silent Mutation CTG,TTG L497L XP_006724506.2
XM_011546178.2 1287 Silent Mutation CTG,TTG L308L XP_011544480.1
XM_011546179.2 1287 Silent Mutation CTG,TTG L308L XP_011544481.1

View Full Product Details