Product Details

SNP ID
rs142270401
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54539061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAGAAAATAAAAAGCCAGGTGAAG[A/G]TTCCAAGGGCCACAAGAAGATAAGT
Phenotype
MIM: 300873
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GNL3L PubMed Links

Gene Details

Gene
GNL3L
Gene Name
G protein nucleolar 3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184819.1 180 Missense Mutation GAT,GGT D14G NP_001171748.1
NM_019067.5 180 Missense Mutation GAT,GGT D14G NP_061940.1

View Full Product Details