Product Details

SNP ID
rs142684980
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:72205167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTCCACGCTTTACAAGAGTCTC[A/G]TAGTCATTTGTAGCCTCTGCCGCCT
Phenotype
MIM: 300687 MIM: 300252
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ERCC6L PubMed Links

Gene Details

Gene
ERCC6L
Gene Name
ERCC excision repair 6 like, spindle assembly checkpoint helicase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017669.2 3697 Silent Mutation TAC,TAT Y1200Y NP_060139.2
Gene
PIN4
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170747.1 3697 Intron NP_001164218.1
NM_006223.3 3697 Intron NP_006214.2

View Full Product Details