Product Details

SNP ID
rs142871346
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:119870608 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTAATGGGAATTGCATCCTCAT[C/T]GGGGTCTTCTGGCAAGTCGGAAGCA
Phenotype
MIM: 300078 MIM: 300951
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
NDUFA1 PubMed Links

Gene Details

Gene
NDUFA1
Gene Name
NADH:ubiquinone oxidoreductase subunit A1
There are no transcripts associated with this gene.

Gene
RNF113A
Gene Name
ring finger protein 113A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006978.2 1221 Missense Mutation AAT,GAT N336D NP_008909.1

View Full Product Details