Product Details

SNP ID
rs142893082
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:103825458 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAATGCTCAGGGTCCTTCACATC[C/T]GCATAGTAAATAAATTCTTTCTGCC
Phenotype
MIM: 300285
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB9B PubMed Links

Gene Details

Gene
RAB9B
Gene Name
RAB9B, member RAS oncogene family
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016370.2 666 Silent Mutation GCA,GCG A109A NP_057454.1

View Full Product Details