Product Details

SNP ID
rs143149392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:126551508 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATAGAAGAGCAGGGAGCCCTGACC[A/G]GTGCCCACAGTGATGATGTGGCGGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCAF12L1 PubMed Links

Gene Details

Gene
DCAF12L1
Gene Name
DDB1 and CUL4 associated factor 12 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178470.4 1352 Silent Mutation ACA,ACG T367T NP_848565.2

View Full Product Details