Product Details

SNP ID
rs143243756
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15391765 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTAGACTTTAGCTTGTCCTCATCCT[C/T]CTGCATGCCATGTCCAGCTCTTCTT
Phenotype
MIM: 300091 MIM: 300931
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FIGF PubMed Links

Gene Details

Gene
FIGF
Gene Name
c-fos induced growth factor
There are no transcripts associated with this gene.

Gene
PIR
Gene Name
pirin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018109.2 Intron NP_001018119.1
NM_003662.3 Intron NP_003653.1
Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details