Product Details

SNP ID
rs143362612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153649365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGGGCAGCCTGTGCCTGGGCTC[C/T]GACTGCTCTGATGCGGAATCCGAGG
Phenotype
MIM: 300134
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DUSP9 PubMed Links

Gene Details

Gene
DUSP9
Gene Name
dual specificity phosphatase 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318503.1 624 Silent Mutation TCC,TCT S169S NP_001305432.1
NM_001395.3 624 Silent Mutation TCC,TCT S169S NP_001386.1
XM_011531123.1 624 Silent Mutation TCC,TCT S200S XP_011529425.1
XM_011531124.1 624 Silent Mutation TCC,TCT S169S XP_011529426.1

View Full Product Details