Product Details

SNP ID
rs143510014
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:15355270 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTAACAGGCACTAATTCAGGTACT[C/G]ATGTCAAAGGCACTGATATCTCAAA
Phenotype
MIM: 300091
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FIGF PubMed Links

Gene Details

Gene
FIGF
Gene Name
c-fos induced growth factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004469.4 988 Nonsense Mutation TCA,TGA S174* NP_004460.1
Gene
PIR-FIGF
Gene Name
PIR-FIGF readthrough
There are no transcripts associated with this gene.

View Full Product Details