Product Details
- SNP ID
-
rs143554153
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:48512706 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GTACTTCATCCCCCTCAACGGTGAC[C/T]GCCTCCTTCGCAAGTGAGCACAGCC
- Phenotype
-
MIM: 300205
MIM: 300651
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EBP
PubMed Links
Gene Details
- Gene
- EBP
- Gene Name
- emopamil binding protein (sterol isomerase)
There are no transcripts associated with this gene.
- Gene
- LOC101927635
- Gene Name
- uncharacterized LOC101927635
There are no transcripts associated with this gene.
- Gene
- PORCN
- Gene Name
- porcupine homolog (Drosophila)
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282167.1 |
992 |
Missense Mutation |
CGC,TGC |
R154C |
NP_001269096.1 |
NM_022825.3 |
992 |
Missense Mutation |
CGC,TGC |
R225C |
NP_073736.2 |
NM_203473.2 |
992 |
Missense Mutation |
CGC,TGC |
R225C |
NP_982299.1 |
NM_203474.1 |
992 |
Missense Mutation |
CGC,TGC |
R225C |
NP_982300.1 |
NM_203475.2 |
992 |
Missense Mutation |
CGC,TGC |
R225C |
NP_982301.1 |
XM_005272635.2 |
992 |
Missense Mutation |
CGC,TGC |
R338C |
XP_005272692.1 |
XM_005272636.2 |
992 |
Missense Mutation |
CGC,TGC |
R338C |
XP_005272693.1 |
XM_005272637.1 |
992 |
Missense Mutation |
CGC,TGC |
R309C |
XP_005272694.1 |
XM_006724545.3 |
992 |
Missense Mutation |
CGC,TGC |
R242C |
XP_006724608.1 |
XM_006724546.3 |
992 |
Missense Mutation |
CGC,TGC |
R225C |
XP_006724609.1 |
XM_006724547.1 |
992 |
Missense Mutation |
CGC,TGC |
R154C |
XP_006724610.1 |
XM_011543948.2 |
992 |
Missense Mutation |
CGC,TGC |
R154C |
XP_011542250.1 |
XM_017029735.1 |
992 |
Missense Mutation |
CGC,TGC |
R338C |
XP_016885224.1 |
XM_017029736.1 |
992 |
Missense Mutation |
CGC,TGC |
R154C |
XP_016885225.1 |
View Full Product Details