Product Details
- SNP ID
-
hCV175413896
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:155033410 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACGTATTACTTCTTGTAGGAAACTT[C/T]GAGGGAAATTTTGAGTCACTGGACC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FUNDC2
PubMed Links
Gene Details
- Gene
- FUNDC2
- Gene Name
- FUN14 domain containing 2
There are no transcripts associated with this gene.
View Full Product Details