Product Details

SNP ID
rs143689285
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:48805666 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAAAAGGAAGAGCTGATGTTGGT[C/T]CACAGGTGAAGGCTTGGGAGCCTTG
Phenotype
MIM: 300272
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HDAC6 PubMed Links

Gene Details

Gene
HDAC6
Gene Name
histone deacetylase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321225.1 719 Silent Mutation GTC,GTT V158V NP_001308154.1
NM_001321226.1 719 Silent Mutation GTC,GTT V144V NP_001308155.1
NM_001321227.1 719 Silent Mutation GTC,GTT V144V NP_001308156.1
NM_001321228.1 719 Silent Mutation GTC,GTT V144V NP_001308157.1
NM_001321229.1 719 Silent Mutation GTC,GTT V144V NP_001308158.1
NM_001321230.1 719 Silent Mutation GTC,GTT V144V NP_001308159.1
NM_001321231.1 719 Silent Mutation GTC,GTT V144V NP_001308160.1
NM_006044.3 719 Silent Mutation GTC,GTT V144V NP_006035.2
XM_017029187.1 719 Silent Mutation GTC,GTT V158V XP_016884676.1

View Full Product Details