Product Details

SNP ID
rs143812963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:21839766 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGGTGTCGCTGGTGGTGGTGGTGG[G/T]GGGTGGCTGGACTGTCGTCTACCTG
Phenotype
MIM: 300294
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MBTPS2 PubMed Links

Gene Details

Gene
MBTPS2
Gene Name
membrane bound transcription factor peptidase, site 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015884.3 229 Missense Mutation GGG,GTG G11V NP_056968.1

View Full Product Details