Product Details

SNP ID
rs144058777
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:88753719 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCCATTCCCAGAAAATTGGTCT[A/C]TCACAAGCCCTTAAATGATAGATCA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CPXCR1 PubMed Links

Gene Details

Gene
CPXCR1
Gene Name
CPX chromosome region, candidate 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001184771.1 564 Missense Mutation TAT,TCT Y102S NP_001171700.1
NM_033048.5 564 Missense Mutation TAT,TCT Y102S NP_149037.4

View Full Product Details