Product Details

SNP ID
rs144106353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71368115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGACATCAATGAGGTGGCAGAAGA[C/T]GAAAGCCGAAGATACCAGCAGACGA
Phenotype
MIM: 313650
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TAF1 PubMed Links

Gene Details

Gene
TAF1
Gene Name
TATA-box binding protein associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286074.1 452 Intron NP_001273003.1
NM_004606.4 452 Intron NP_004597.2
NM_138923.3 452 Intron NP_620278.1
XM_005262295.1 452 Intron XP_005262352.1
XM_005262296.1 452 Intron XP_005262353.1
XM_005262297.4 452 Silent Mutation GAC,GAT D119D XP_005262354.1
XM_005262300.2 452 Intron XP_005262357.1
XM_006724682.3 452 Intron XP_006724745.1

View Full Product Details