Product Details

SNP ID
rs144336465
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30665035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCTCTTTTATAAGAGCTAGATGC[A/G]TAGCCACACTTCCCTTCAAAGGAAT
Phenotype
MIM: 300474
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GK PubMed Links

Gene Details

Gene
GK
Gene Name
glycerol kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000167.5 Intron NP_000158.1
NM_001128127.2 Intron NP_001121599.1
NM_001205019.1 Intron NP_001191948.1
NM_203391.3 Intron NP_976325.1
XM_005274488.4 Intron XP_005274545.1
XM_006724483.2 Intron XP_006724546.1
XM_006724484.2 Intron XP_006724547.1
XM_006724485.2 Intron XP_006724548.1
XM_006724486.3 Intron XP_006724549.1
XM_011545491.2 Intron XP_011543793.1
XM_011545492.1 Intron XP_011543794.1
XM_011545493.2 Intron XP_011543795.1
XM_011545494.2 Intron XP_011543796.1
XM_017029409.1 Intron XP_016884898.1
XM_017029410.1 Intron XP_016884899.1
XM_017029411.1 Intron XP_016884900.1
XM_017029412.1 Intron XP_016884901.1

View Full Product Details