Product Details

SNP ID
rs144428691
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:131544624 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCTGTGAGATTCTCTCCCTCA[A/T]TAAGCTGACCTGTTCTGATACCAGC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
OR13H1 PubMed Links

Gene Details

Gene
OR13H1
Gene Name
olfactory receptor family 13 subfamily H member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004486.1 551 Missense Mutation AAT,ATT N184I NP_001004486.1

View Full Product Details