Product Details

SNP ID
rs144502161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:81115003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTCTCTTTTGCATCTTCTCCTTT[C/G]TCATTTCCATTTTTATCCTCTTTTC
Phenotype
MIM: 300385
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HMGN5 PubMed Links

Gene Details

Gene
HMGN5
Gene Name
high mobility group nucleosome binding domain 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030763.2 654 Missense Mutation GAC,GAG D165E NP_110390.1
XM_006724688.3 654 Missense Mutation GAC,GAG D166E XP_006724751.1
XM_017029839.1 654 Missense Mutation GAC,GAG D156E XP_016885328.1
XM_017029840.1 654 Missense Mutation GAC,GAG D155E XP_016885329.1

View Full Product Details