Product Details

SNP ID
rs144513627
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153767131 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGACAGCCCTGACTGCGTGCCCTTC[C/T]GTGACCCAGCCGAGTGCCCACAGGC
Phenotype
MIM: 300214
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLXNB3 PubMed Links

Gene Details

Gene
PLXNB3
Gene Name
plexin B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163257.1 478 Missense Mutation CGT,TGT R125C NP_001156729.1
NM_005393.2 478 Missense Mutation CGT,TGT R102C NP_005384.2

View Full Product Details