Product Details

SNP ID
rs144634143
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:80373647 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGCTATATATCTATTCCTCTTG[A/G]TTAATCCTTCTCTCCTGATCTCCCA
Phenotype
MIM: 300976
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM46D PubMed Links

Gene Details

Gene
FAM46D
Gene Name
family with sequence similarity 46 member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170574.1 Intron NP_001164045.1
NM_152630.4 Intron NP_689843.1

View Full Product Details