Product Details
- SNP ID
-
rs144925686
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:103079862 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTGTCGATCTCCAGAGTCATAGATC[A/G]AGTAATACCTGTTGGAGACCAGTGA
- Phenotype
-
MIM: 300316
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
NXF3
PubMed Links
Gene Details
- Gene
- NXF3
- Gene Name
- nuclear RNA export factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022052.1 |
1132 |
Missense Mutation |
TCG,TTG |
S354L |
NP_071335.1 |
View Full Product Details