Product Details

SNP ID
rs145061343
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:74422049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGAGGAAAAGGAAAAAAATCGC[C/G]AAGTGGAGTTCCAAGCAGGTGAGTG
Phenotype
MIM: 300095
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SLC16A2 PubMed Links

Gene Details

Gene
SLC16A2
Gene Name
solute carrier family 16 member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006517.4 557 Missense Mutation CAA,GAA Q138E NP_006508.2

View Full Product Details