Product Details

SNP ID
rs145090274
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:80017988 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTTCGCTTCCAATCCTCCCCATG[A/G]GAAGAACACAGCGAACACAGATAAG
Phenotype
MIM: 300307
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TBX22 PubMed Links

Gene Details

Gene
TBX22
Gene Name
T-box 22
There are no transcripts associated with this gene.

View Full Product Details