Product Details

SNP ID
rs145282692
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71108289 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGCGTTCTCACCGAAAAGTTCCC[A/G]TGGTATTCAGTAACAAGATCCTCTA
Phenotype
MIM: 300033 MIM: 308380
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
There are no transcripts associated with this gene.

Gene
FOXO4
Gene Name
forkhead box O4
There are no transcripts associated with this gene.

Gene
IL2RG
Gene Name
interleukin 2 receptor subunit gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000206.2 1004 Silent Mutation CAC,CAT H304H NP_000197.1

View Full Product Details