Product Details

SNP ID
hCV175466445
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153941493 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTCCCACCTGGGTGGGGCAGAA[C/G]TTATCAGCATCCTGGAAGTAAAAGA
Phenotype
MIM: 300019 MIM: 300013 MIM: 312420
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HCFC1 PubMed Links

Gene Details

Gene
HCFC1
Gene Name
host cell factor C1
There are no transcripts associated with this gene.

Gene
NAA10
Gene Name
N(alpha)-acetyltransferase 10, NatA catalytic subunit
There are no transcripts associated with this gene.

Gene
RENBP
Gene Name
renin binding protein
There are no transcripts associated with this gene.

View Full Product Details